Learn More
Waters Corp ACQUITY - Xselect HSS C18 SB Method Transfer Kit, 100Å, 1.8 - 3.5 µm, 2.1 - 4.6 mm X 50 - 100mm, 2/pk
Catalog No. 50791274 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Method transfer kits are designed to preserve the integrity of a separation as it is transferred between UPLC and HPLC platforms. Based on the concept of maintaining column length [L] to particle size [dp] ratio [L/dp], these kits provide an ACQUITY UPLC column with an HPLC column of equivalent selectivity and resolving power. Using the ACQUITY UPLC columns calculator, methods can be fully transferred from HPLC to UPLC or from UPLC to HPLC.
Catalog No. 50-791-274 Supplier Waters Corp Supplier No. 186004970
Add to Cart
Add to Cart
Method transfer kits are designed to preserve the integrity of a separation as it is transferred between UPLC and HPLC platforms. Based on the concept of maintaining column length [L] to particle size [dp] ratio [L/dp], these kits provide an ACQUITY UPLC column with an HPLC column of equivalent selectivity and resolving power. Using the ACQUITY UPLC columns calculator, methods can be fully transferred from HPLC to UPLC or from UPLC to HPLC.