Learn More
Waters Corp XSelect HSS C18 SB Column, 100Å, 3.5 µm, 3 mm X 50 mm, 1/pk
Catalog No. 50466065 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HSS C18 SB (Selectivity for Bases) is an unendcapped, low-coverage C18 bonded-phase column that was designed specifically to provide increased silanophilic interactions. This results in an alternate selectivity compared to a high-coverage fully-endcapped C18 column, particularly for basic compounds.
Catalog No. 50-466-065 Supplier Waters Corp Supplier No. 186004747
The HSS C18 SB (Selectivity for Bases) is an unendcapped, low-coverage C18 bonded-phase column that was designed specifically to provide increased silanophilic interactions. This results in an alternate selectivity compared to a high-coverage fully-endcapped C18 column, particularly for basic compounds.