Learn More
Waters Corp Waters XSelect HSS C18 SB Column, 100Å, 3.5 μm, 3 mm × 150 mm
Supplier:  Waters Corp 186004744
Encompass_Preferred
The HSS C18 SB (Selectivity for Bases) column is an unendcapped, low-coverage C18 bonded-phase. It offers increased silanophilic interactions and alternate selectivity, especially for basic compounds. Built with HSS—High Strength Silica technology, it withstands UPLC pressures, making it scalable for UPLC separations and enhancing detection of polar compounds.
  • Unendcapped, low-coverage C18 bonded-phase.
  • Increased silanophilic interactions and alternate selectivity for basic and polar compounds.
  • HSS (High Strength Silica) technology.
  • UPLC pressure compatibility and scalability.
  • Recommended pH range: 2 to 8.
  • Withstands pressure up to 6000 psi (415 bar) and temperature from 20°C to 45°C.
Catalog No. 50-466-062