Learn More
Waters Corp Waters XSelect HSS C18 Sb Column, 3.5 μm, 100 Å, 4.6 mm x 100 mm
Catalog No. 50466066 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect HSS C18 SB Column is an unendcapped, low-coverage C18 bonded-phase column offering alternate selectivity for basic compounds. Utilizing HSS—High Strength Silica technology, it enhances chromatographic performance and withstands UPLC pressures for scalable separations.
  • Unendcapped, low-coverage C18 bonded-phase for increased silanophilic interactions.
  • Provides alternate selectivity, especially for basic compounds.
  • Utilizes high strength silica (HSS) technology.
  • Improves chromatographic performance of BEH and CSH particles.
  • Withstands UPLC pressures for scalability and transferability.
  • Offers different selectivity and higher polar compound retention.
Catalog No. 50-466-066 Supplier Waters Corp Supplier No. 186004748
Add to Cart
Add to Cart
The XSelect HSS C18 SB Column is an unendcapped, low-coverage C18 bonded-phase column offering alternate selectivity for basic compounds. Utilizing HSS—High Strength Silica technology, it enhances chromatographic performance and withstands UPLC pressures for scalable separations.
  • Unendcapped, low-coverage C18 bonded-phase for increased silanophilic interactions.
  • Provides alternate selectivity, especially for basic compounds.
  • Utilizes high strength silica (HSS) technology.
  • Improves chromatographic performance of BEH and CSH particles.
  • Withstands UPLC pressures for scalability and transferability.
  • Offers different selectivity and higher polar compound retention.