Learn More
Waters Corp XSelect HSS T3 Column, 3.5 μm, 3 mm, 100 mm, 100 Å
Supplier:  Waters Corp 186004780
Encompass_Preferred
The HSS T3 column is a fully endcapped, low-coverage C18 bonded-phase column designed to retain extremely polar compounds by reversed-phase in high aqueous mobile phases. High Strength Silica (HSS) Technology complements chromatographic performance and scalability, offering higher silanophilicity for increased polar retention and selectivity.
  • Recommended for separations of both polar and nonpolar compounds.
  • Features a low-coverage C18 phase compatible with 100% aqueous mobile phase.
  • Offers ultra-low MS bleed and full scalability to UPLC separations.
  • Designed for supreme reproducibility and transferability across systems.
  • Enables higher resolution for complex samples, facilitating isolation and quantification.
  • Combines efficiency and selectivity to enhance resolution of very complex samples.
Catalog No. 50-466-097