Learn More
Waters Corp Waters Column, 100Å, 3.5 μm, 3 mm X 150 mm
Catalog No. 50466098 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HSS T3 column is a fully endcapped, low-coverage C18 bonded-phase column designed for reversed-phase retention of highly polar compounds in high aqueous mobile phases. Utilizing High Strength Silica (HSS) Technology, it offers supreme reproducibility, transferability, and seamless scalability for high-resolution separations.
  • Supreme reproducibility and transferability for consistent results.
  • Seamless scalability for various analytical needs.
  • Higher resolution separations for complex samples.
  • Enhanced efficiency and selectivity for better resolution.
  • HSS technology provides higher silanophilicity and increased polar compound retention.
  • High Strength Silica (HSS) technology withstands UPLC pressures.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase.
  • Ultra-low MS bleed; fully scalable and transferable to UPLC separations.
Catalog No. 50-466-098 Supplier Waters Corp Supplier No. 186004781
This HSS T3 column is a fully endcapped, low-coverage C18 bonded-phase column designed for reversed-phase retention of highly polar compounds in high aqueous mobile phases. Utilizing High Strength Silica (HSS) Technology, it offers supreme reproducibility, transferability, and seamless scalability for high-resolution separations.
  • Supreme reproducibility and transferability for consistent results.
  • Seamless scalability for various analytical needs.
  • Higher resolution separations for complex samples.
  • Enhanced efficiency and selectivity for better resolution.
  • HSS technology provides higher silanophilicity and increased polar compound retention.
  • High Strength Silica (HSS) technology withstands UPLC pressures.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase.
  • Ultra-low MS bleed; fully scalable and transferable to UPLC separations.