Learn More
Waters Corp XSelect HSS T3 Column, 100Å, 3.5 µm, 3 mm X 30 mm, 1/pk
Catalog No. 50466100 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HSS T3 column is a fully endcapped, low-coverage C18 bonded-phase column designed to retain extremely polar compounds by reversed-phase in high aqueous mobile phases.
Catalog No. 50-466-100 Supplier Waters Corp Supplier No. 186004783
The HSS T3 column is a fully endcapped, low-coverage C18 bonded-phase column designed to retain extremely polar compounds by reversed-phase in high aqueous mobile phases.