Learn More
Waters Corp XSelect Column, HSS T3, 100 Å, 5 μm, 4.6 mm X 150 mm
Supplier:  Waters Corp 186004791
Encompass_Preferred
XSelect HSS T3 Columns redefine modern HPLC technology, leveraging High Strength Silica and T3 processes for unique selectivity, high analyte loading, operational stability, and full scalability. Designed for UPLC with high-pressure tolerance, they feature a unique silica substrate and optimal ligand bonding for high aqueous mobile phases. The proprietary T3 end-capping enhances retention, performance, lifetime, peak shape, and stability. Each column is individually tested, manufactured in a cGMP, ISO 9001:2000 certified plant, and includes a certificate of batch analysis and a performance chromatogram.
  • Fully endcapped, low-coverage C18 bonded-phase column.
  • Designed to retain extremely polar compounds by reversed-phase in high aqueous mobile phases.
  • High pressure tolerance for UPLC applications.
Catalog No. 50-466-108