Learn More
MSC OSG Thread Plug Gage, 9/16-18 UN(J)F, 3B, TiN, 1 ea
Catalog No. 5000514619
Encompass
This thread limit gage is a high-precision inspection tool designed for checking the accuracy of 9/16-18 UN(J)F class 3B threaded parts. Constructed from high-speed steel with a titanium nitride (TiN) coating, this double-ended gage offers superior hardness and extended tool life in demanding inspection environments.
  • Manufactured from durable high-speed steel
  • Features a titanium nitride (TiN) coating for enhanced wear resistance
  • Double-ended design with go and no-go components
  • Class 3B thread fit accuracy
  • Hardness rating of 64 Rc
  • Includes short form certification
Catalog No. 50-005-14619 Supplier MSC Supplier No. 72642036
This thread limit gage is a high-precision inspection tool designed for checking the accuracy of 9/16-18 UN(J)F class 3B threaded parts. Constructed from high-speed steel with a titanium nitride (TiN) coating, this double-ended gage offers superior hardness and extended tool life in demanding inspection environments.
  • Manufactured from durable high-speed steel
  • Features a titanium nitride (TiN) coating for enhanced wear resistance
  • Double-ended design with go and no-go components
  • Class 3B thread fit accuracy
  • Hardness rating of 64 Rc
  • Includes short form certification