Learn More
Electron Microscopy Sciences HistoPro 200 Parafin Infiltration Processor
SDP
Catalog No. 501926624
Encompass_Preferred
HistoPro 200 offers unique reagent / paraffin infiltration capability with programmable processing time, temperature, agitation and vacuum in an easy to use, compact, cost conscious package. Holds up to 100 cassettes. 2.0 liter reagent / paraffin capacity. Compact footprint: only 9" wide x 20" depth. Programmable processing time, programmable processing temperature, and programmable vacuum cycle.
Catalog No. 50-192-6624 Supplier Electron Microscopy Sciences Supplier No. 6259610
HistoPro 200 offers unique reagent / paraffin infiltration capability with programmable processing time, temperature, agitation and vacuum in an easy to use, compact, cost conscious package. Holds up to 100 cassettes. 2.0 liter reagent / paraffin capacity. Compact footprint: only 9" wide x 20" depth. Programmable processing time, programmable processing temperature, and programmable vacuum cycle.