Learn More
Creative Biomart Recombinant HSV-1 gD Antigen
Supplier:  Creative Biomart HSV234
Encompass_Preferred
The E. coli derived recombinant protein contains the HSV-1 gD (84-175) immunodominant region.
Catalog No. 50-208-4612