Learn More
Enzo Life Sciences HSV-1 native antigen (200µl)
SDP
Supplier:  Enzo Life Sciences ALX6301260200
Encompass_Preferred
Alternative name: Herpes simplex virus type 1 antigen. Purity: ≥80%. Appearance: Cloudy liquid. Formulation: Liquid. In PBS containing 0.1% Triton X-100. Source: From herpes simplex virus type 1. Inactivated by 0.1% Triton X-100. Long Term Storage: -80°C. Applications: ELISA Positive control, Antibody capture.
Catalog No. 50-200-9652