Learn More
Creative Biomart Recombinant HSV-2 gB Antigen, His-tagged
Supplier:  Creative Biomart HSV236
Encompass_Preferred
The E. coli derived HSV-2 gB recombinant protein is fused to a Six histidine tag at C-terminus and has a MW of 82 kDa (pI 8.35).
Catalog No. 50-208-4614
 Restricted Item