Learn More
Creative Biomart Recombinant HSV-2 gD Antigen, GST-tagged
Supplier:  Creative Biomart HSV237
Encompass_Preferred
The E. coli derived 39.7 kDa recombinant protein contains the HSV-2 gD immunodominant regions 266-394 amino acids, fused with 26 kDa GST-tag.
Catalog No. 50-208-4615