Learn More
ACROBiosystems HSV-2 (strain 333) Glycoprotein B (gB), His Tag (MALS verified)
Catalog No. 502719587
Encompass_Preferred
Envelope glycoprotein that forms spikes at the surface of virion envelope. Essential for the initial attachment to heparan sulfate moieties of the host cell surface proteoglycans. Involved in fusion of viral and cellular membranes leading to virus entry into the host cell. Following initial binding to its host receptors, membrane fusion is mediated by the fusion machinery composed at least of gB and the heterodimer gH/gL. May be involved in the fusion between the virion envelope and the outer nuclear membrane during virion egress. Also plays a role, together with gK, in virus-induced cell-to-cell fusion (syncytia formation).
Catalog No. 50-271-9587 Supplier ACROBiosystems Supplier No. GLBH52H350UG
Envelope glycoprotein that forms spikes at the surface of virion envelope. Essential for the initial attachment to heparan sulfate moieties of the host cell surface proteoglycans. Involved in fusion of viral and cellular membranes leading to virus entry into the host cell. Following initial binding to its host receptors, membrane fusion is mediated by the fusion machinery composed at least of gB and the heterodimer gH/gL. May be involved in the fusion between the virion envelope and the outer nuclear membrane during virion egress. Also plays a role, together with gK, in virus-induced cell-to-cell fusion (syncytia formation).