Learn More
Drg International Inc DRG HSV-2 IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3487R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Herpes simplex Virus Type 2 (HSV-2) in human serum and plasma. The DRG HSV 2 IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) for Research Use Only. Microtiter wells, as a solid phase, are coated with native gG2 protein of HSV-2.  Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7575