Learn More
Enzo Life Sciences HSV-2 native antigen (200µl)
SDP
Supplier:  Enzo Life Sciences ALX6301270200
Encompass_Preferred
Alternative name: Herpes simplex virus type 2 antigen. Purity: ≥80%. Formulation: Liquid. In PBS containing 0.1% Triton X-100. Source: From herpes simplex virus type 2, strain G. Inactivated by 0.1% Triton X-100. Long Term Storage: -80°C. Applications: ELISA Positive control, Antibody capture.
Catalog No. 50-200-9653