Learn More
CELL MARQUE CORPORATION HSV CONTROLS
SDP
Catalog No. NC1995947
Encompass
Herpes Simplex Virus I & II positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-Herpes Simplex Virus I & II antibody. Each pack contains 5 paraffin embedded tissue sections. The preserved tissue sections of approximately 2-4 µm are embedded in paraffin and heat fixed on positively charged slides.
Catalog No. NC1995947 Supplier CELL MARQUE CORPORATION Supplier No. 214S
Herpes Simplex Virus I & II positive control slides are intended for use as positive controls for immunohistochemical (IHC) staining using an anti-Herpes Simplex Virus I & II antibody. Each pack contains 5 paraffin embedded tissue sections. The preserved tissue sections of approximately 2-4 µm are embedded in paraffin and heat fixed on positively charged slides.