Learn More
Diagnostic Biosystems HSV I Herpes Simplex Virus Type I (HSV1)
Catalog No. 502407578
Encompass_Preferred
This antibody reacts with HSV type I specific antigens and with antigens common to HSV type I and II virus. It reacts with all the major glycoproteins present in the viral envelope as well as with at least one core protein.
Catalog No. 50-240-7578 Supplier Diagnostic Biosystems Supplier No. PDR032
This antibody reacts with HSV type I specific antigens and with antigens common to HSV type I and II virus. It reacts with all the major glycoproteins present in the viral envelope as well as with at least one core protein.