Learn More
Sino Biological HSV 2 (strain 333) gD Protein (ECD, His Tag)(MALS-verified)
Supplier:  Sino Biological 40936V08H100
Encompass_Preferred
A DNA sequence encoding the HSV 2 (strain 333 (HSV2)) gD (translated amino acids of P03172) (Lys26-Thr310) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2743