Learn More
Aviva Systems Biology Corp HSZFP36 Antibody - N-terminal region (ARP50464_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50464_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HSZFP36. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSZFP36. Alternate names: HSZFP36ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53632