Learn More
Aviva Systems Biology Corp HSZFP36 Antibody - middle region (ARP32899_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32899_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSZFP36. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSZFP36. Alternate names: HSZFP36ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-54811