Learn More
Cayman Chemical HT-2 Toxin-13C22, Purity: ≥98%, CAS: 1486469-92-4, 1.2 mL
SDP
Catalog No. 502991582
Encompass_Preferred
An internal standard for the quantification of HT-2 toxin by GC- or LC-MS
Catalog No. 50-299-1582 Supplier Cayman Chemical Supplier No. 312991.2 ML
An internal standard for the quantification of HT-2 toxin by GC- or LC-MS