Learn More
Enzo Life Sciences HT-2 Toxin (1mg). CAS: 26934-87-2
SDP
Catalog No. 502009638
Encompass_Preferred
Trichothecene group mycotoxin. Induces apoptosis Alternative name: 15-Acetoxy-3alpha,4beta-dihydroxy-8alpha-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol, DMSO or ethyl acetate. Slightly soluble in petroleum ether; poorly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9638 Supplier Enzo Life Sciences Supplier No. ALX630113M001
Add to Cart
Add to Cart
Trichothecene group mycotoxin. Induces apoptosis Alternative name: 15-Acetoxy-3alpha,4beta-dihydroxy-8alpha-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol, DMSO or ethyl acetate. Slightly soluble in petroleum ether; poorly soluble in water. Long Term Storage: -20°C.