Learn More
Waters Corp HT Diverter Valve Pod, Mass Spectrometry, Replacement Part
Catalog No. 502392791 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HT Diverter Valve Pod is a replacement component specifically designed for the diverter valve assembly found in a range of Mass Spectrometers. This includes models such as the Synapt G2, Synapt G2-S/Si, Xevo QTOF, Xevo G2, Xevo G2-S/XS, Vion, Xevo TQ MS, Xevo TQ-S, and Xevo TQ-XS, ensuring their continued operation and performance.
  • Replacement component for diverter valve assemblies.
  • Compatible with multiple mass spectrometer systems.
  • Supports proper instrument functionality.
  • Classified as a spare part kit for maintenance.
Catalog No. 50-239-2791 Supplier Waters Corp Supplier No. 700009461
The HT Diverter Valve Pod is a replacement component specifically designed for the diverter valve assembly found in a range of Mass Spectrometers. This includes models such as the Synapt G2, Synapt G2-S/Si, Xevo QTOF, Xevo G2, Xevo G2-S/XS, Vion, Xevo TQ MS, Xevo TQ-S, and Xevo TQ-XS, ensuring their continued operation and performance.
  • Replacement component for diverter valve assemblies.
  • Compatible with multiple mass spectrometer systems.
  • Supports proper instrument functionality.
  • Classified as a spare part kit for maintenance.