Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup

Catalog No. p-7138019
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
1000 reactions
5000 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
78-595-5000RX 5000 reactions
78-595-1000RX 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 78-595-5000RX Supplier Applied Biosystems™ Supplier No. 785955000RXN
Only null left
Add to Cart
Add to Cart
Request a Quote

HT ExoSAP-IT Fast High-Throughput PCR Product Clean-up, with its shortened 14 minute protocol, is a highly stable, customized formulation of the original ExoSAP-IT reagent

  • 14 minute protocol is half the time of standard protocols
  • One-tube PCR clean-up - add HT ExoSAP-IT Fast reagent directly to PCR product
  • 100% sample recovery - no loss of PCR products regardless of the fragment size
  • Stable at 4°C for one month
  • Removes excess primers and dNTPs - does not interfere with downstream applications sequencing, SNP analysis, single base extension, fragment analysis
  • It is ideal for high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes
  • Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Format 8 Tube Strip, 96-well Plate, Vial
Final Product Type Fast High-Throughput PCR Product Clean-up
For Use With (Application) For high-throughput PCR product clean-up using liquid handling platforms and multi-channel pipettes
No. of Reactions 5000 reactions
Product Type HT ExoSAP-IT™ Fast High-Throughput PCR Product Clean-up
Quantity Each
Test Time 14 min

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.