Learn More
Wilmad Labglass HT MNTL BENCH TOP HEMI 12L
Catalog No. 50801201
Encompass
Heating Mantle, Hemispherical, Aluminum Housing. Size: 12000mL; Maximum Temperature: 450degreeC; Watts: 2x650; Volts: 115. Bench top, aluminum-housed heating mantle for round bottom flasks. Supplied w/ 4 foot long 3-Wire type SJT cord.
Catalog No. 50-801-201 Supplier Wilmad Labglass Supplier No. LG8875122
Heating Mantle, Hemispherical, Aluminum Housing. Size: 12000mL; Maximum Temperature: 450degreeC; Watts: 2x650; Volts: 115. Bench top, aluminum-housed heating mantle for round bottom flasks. Supplied w/ 4 foot long 3-Wire type SJT cord.