Learn More
Wilmad Labglass HT MNTL BENCH TOP HEMI 3L
Catalog No. 50801199
Encompass
Heating Mantle, Hemispherical, Aluminum Housing. Size: 3000mL; Maximum Temperature: 450degreeC; Watts: 500; Volts: 115. Bench top, aluminum-housed heating mantle for round bottom flasks. Supplied w/ 4 foot long 3-Wire type SJT cord.
Catalog No. 50-801-199 Supplier Wilmad Labglass Supplier No. LG8875118
Heating Mantle, Hemispherical, Aluminum Housing. Size: 3000mL; Maximum Temperature: 450degreeC; Watts: 500; Volts: 115. Bench top, aluminum-housed heating mantle for round bottom flasks. Supplied w/ 4 foot long 3-Wire type SJT cord.