Learn More
Wilmad Labglass HT MNTL BENCH TOP HEMI 5L
Catalog No. 50801200
Encompass
Heating Mantle, Hemispherical, Aluminum Housing. Size: 5000mL; Maximum Temperature: 450degreeC; Watts: 600; Volts: 115. Bench top, aluminum-housed heating mantle for round bottom flasks. Supplied w/ 4 foot long 3-Wire type SJT cord.
Catalog No. 50-801-200 Supplier Wilmad Labglass Supplier No. LG8875120
Heating Mantle, Hemispherical, Aluminum Housing. Size: 5000mL; Maximum Temperature: 450degreeC; Watts: 600; Volts: 115. Bench top, aluminum-housed heating mantle for round bottom flasks. Supplied w/ 4 foot long 3-Wire type SJT cord.