Learn More
Wilmad Labglass HT MNTL HEMI 100ML
Catalog No. 50801160
Encompass
Heating Mantle, Hemispherical. Size: 100mL. Watts: 80. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.
Catalog No. 50-801-160 Supplier Wilmad Labglass Supplier No. LG8800102
Heating Mantle, Hemispherical. Size: 100mL. Watts: 80. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.