Learn More
Wilmad Labglass HT MNTL HEMI 250ML
Catalog No. 50801163
Encompass
Heating Mantle, Hemispherical. Size: 250mL. Watts: 180. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.
Catalog No. 50-801-163 Supplier Wilmad Labglass Supplier No. LG8800108
Heating Mantle, Hemispherical. Size: 250mL. Watts: 180. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.