Learn More
Wilmad Labglass HT MNTL HEMI 5000ML
Catalog No. 50801090
Encompass
Heating Mantle, Hemispherical. Size: 5000mL. Watts: 600. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.
Catalog No. 50-801-090 Supplier Wilmad Labglass Supplier No. LG8800120
Heating Mantle, Hemispherical. Size: 5000mL. Watts: 600. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.