Learn More
Wilmad Labglass HT MNTL HEMI 500ML
Catalog No. 50801165
Encompass
Heating Mantle, Hemispherical. Size: 500mL. Watts: 2700. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.
Catalog No. 50-801-165 Supplier Wilmad Labglass Supplier No. LG8800112
Heating Mantle, Hemispherical. Size: 500mL. Watts: 2700. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.