Learn More
Cell Signaling Technology Smad2/3 Control Cell Extracts, HT-1080 Cells, Western Blotting, 2-Piece Set
SDP
Catalog No. NC0583277
Encompass
SMAD2/3 Control Cell Extracts contain total cell extracts from HT-1080 cells, prepared as nonphosphorylated (serum-starved) and phosphorylated (treated with hTGF-β3) forms to function as negative and positive controls, respectively, in western blot analysis. The extracts are supplied in SDS sample buffer and are validated for use in western blotting applications.
  • Provided as a two-vial set of untreated and treated cell lysates
  • Validated for western blot analysis
  • Supplied in SDS sample buffer (62.5 mM Tris-HCl, 2% SDS, 10% glycerol, 50 mM DTT, 0.01% bromophenol blue or phenol red)
  • Ready for use after boiling for 3 minutes
  • Compatible with standard western blotting workflows
Catalog No. NC0583277 Supplier Cell Signaling Technology Supplier No. 12052S
SMAD2/3 Control Cell Extracts contain total cell extracts from HT-1080 cells, prepared as nonphosphorylated (serum-starved) and phosphorylated (treated with hTGF-β3) forms to function as negative and positive controls, respectively, in western blot analysis. The extracts are supplied in SDS sample buffer and are validated for use in western blotting applications.
  • Provided as a two-vial set of untreated and treated cell lysates
  • Validated for western blot analysis
  • Supplied in SDS sample buffer (62.5 mM Tris-HCl, 2% SDS, 10% glycerol, 50 mM DTT, 0.01% bromophenol blue or phenol red)
  • Ready for use after boiling for 3 minutes
  • Compatible with standard western blotting workflows