Learn More
ATCC HT-1376; Bladder Carcinoma; Human (Homo sapiens)
Catalog No. 502382217
Encompass
HT-1376 is an epithelial-like cell isolated from the urinary bladder of a White, 58-year-old, female with carcinoma. This cell line was deposited by S Rasheed and can be used for immuno-oncology research.
Catalog No. 50-238-2217 Supplier ATCC Supplier No. CRL1472
HT-1376 is an epithelial-like cell isolated from the urinary bladder of a White, 58-year-old, female with carcinoma. This cell line was deposited by S Rasheed and can be used for immuno-oncology research.