Learn More
ATCC HT-29; Colon Adenocarcinoma; Human
Supplier:  ATCC HTB38
Encompass
HT-29 is a cell line with epithelial morphology that was isolated in 1964 from a primary tumor obtained from a 44-year-old, White, female patient with colorectal adenocarcinoma. This cell line is a suitable transfection host and has applications in cancer and toxicology research.
Catalog No. 50-238-4466