Learn More
Heska Corporation Heska Element HT5 Lyse Solution, Accurate RBC Dissolving, Ideal for WBC Counting, 90 mL
SDP
Catalog No. NC1989416
Encompass
The Heska Element HT5 LH Lyse Solution is essential for hematology analysis in veterinary settings, aiding in the dissolution of red blood cells for accurate white blood cell counting and hemoglobin measurement. Its nonflammable liquid formulation ensures reliable diagnostics with a transparent appearance and a pH of 2.20. Designed for use with the Heska Element HT5 Auto Hematology Analyzer, this reagent offers high-level performance and precise results, making it a trusted choice for veterinarians.
Catalog No. NC1989416 Supplier Heska Corporation Supplier No. 5225
The Heska Element HT5 LH Lyse Solution is essential for hematology analysis in veterinary settings, aiding in the dissolution of red blood cells for accurate white blood cell counting and hemoglobin measurement. Its nonflammable liquid formulation ensures reliable diagnostics with a transparent appearance and a pH of 2.20. Designed for use with the Heska Element HT5 Auto Hematology Analyzer, this reagent offers high-level performance and precise results, making it a trusted choice for veterinarians.