Learn More
Heska Corporation Heska HT5 Probe Cleaner, Routine Maintenance For Analyzers, 50 mL Pack
SDP
Catalog No. NC1989417
Encompass
The Heska HT5 Probe Cleaner is a crucial cleaning solution specifically designed for Heska Element HT5 and HT5+ hematology analyzers. It features a potent formulation with surfactants and sodium hypochlorite for effective probe cleansing, ensuring precise diagnostic results. Supplied in a convenient 2-bottle pack with 25 mL each, it facilitates routine maintenance and enhances the analyzers' performance. This product stands out with its recommended daily usage to maintain optimal functionality.
Catalog No. NC1989417 Supplier Heska Corporation Supplier No. 5228
The Heska HT5 Probe Cleaner is a crucial cleaning solution specifically designed for Heska Element HT5 and HT5+ hematology analyzers. It features a potent formulation with surfactants and sodium hypochlorite for effective probe cleansing, ensuring precise diagnostic results. Supplied in a convenient 2-bottle pack with 25 mL each, it facilitates routine maintenance and enhances the analyzers' performance. This product stands out with its recommended daily usage to maintain optimal functionality.