Learn More
Medchemexpress LLC hT84.66-M5A 1mg | 1mg
SDP
Catalog No. 5000323330
Encompass_Preferred
hT84 66-M5A is a humanized IgG1 antibody that targets carcinoembryonic antigen (CEA) The isotype control for hT84 66-M5A can refer to Human IgG1 kappa Isotype Control (HY-P99001)[1]
Catalog No. 50-003-23330 Supplier Medchemexpress LLC Supplier No. HYP9910681MG
hT84 66-M5A is a humanized IgG1 antibody that targets carcinoembryonic antigen (CEA) The isotype control for hT84 66-M5A can refer to Human IgG1 kappa Isotype Control (HY-P99001)[1]