Learn More
Gentech Scientific Inc Headspace (10 or 20ml) vial rack
SDP
Catalog No. NC3234943
Encompass_Preferred
Headspace (10 or 20ml) vial rack, Headspace- 42 Position (20ml) headspace vial rack, Headspace Starter Kit: 20ml vials, caps, septa (10pcs), 180 Day Warranty Compatible with all major manufacturer’s GC’s (Agilent, Thermo, Shimadzu and more) GenTech Scientific offers installation, and extended warranty options. Please contact 585-492-1068 with questions.
Catalog No. NC3234943 Supplier Gentech Scientific Inc Supplier No. HTA HT2000H
Add to Cart
Add to Cart
Request a Quote
Headspace (10 or 20ml) vial rack, Headspace- 42 Position (20ml) headspace vial rack, Headspace Starter Kit: 20ml vials, caps, septa (10pcs), 180 Day Warranty Compatible with all major manufacturer’s GC’s (Agilent, Thermo, Shimadzu and more) GenTech Scientific offers installation, and extended warranty options. Please contact 585-492-1068 with questions.