Learn More
Aviva Systems Biology Corp HTATIP Antibody - N-terminal region (ARP32488_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32488_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HTATIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HTATIP. Alternate names: ESA1, HTATIP1, KAT5, PLIP, TIP, TIP60, cPLA2, HTATIPApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54620