Learn More
Aviva Systems Biology Corp HTATIP2 Antibody - N-terminal region (ARP32775_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32775_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HTATIP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HTATIP2. Alternate names: CC3, TIP30, SDR44U1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54756