Learn More
Aviva Systems Biology Corp HTATSF1 Antibody - C-terminal region (ARP31884_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31884_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HTATSF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HTATSF1. Alternate names: TAT-SF1, dJ196E23.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-54352