Learn More
Trajan Scientific PAL Preventative Maintenance Kit - Includes Injection Head, Drawer Magnet, Tension Cords, and More
Catalog No. NC9903343
Encompass_Preferred
Kit, PM, Preventative Maintenance, For Injection head; Includes Drawer Magnet, Short and long tension cords, pin Point oil, torx Driver Set, Ruler, Microlube, Ball Hex: 2.5mm, and Manual
Catalog No. NC9903343 Supplier Trajan Scientific Supplier No. PAL.PMINJKIT
Kit, PM, Preventative Maintenance, For Injection head; Includes Drawer Magnet, Short and long tension cords, pin Point oil, torx Driver Set, Ruler, Microlube, Ball Hex: 2.5mm, and Manual