Learn More
World Precision Instrument HTC Tweezers, Swiss, Straight, Flat
SDP
Supplier:  World Precision Instrument 504617
Encompass
WPI’s Swiss High Temperature Coating (HTC) Thumb Tweezers have undergone a specialized coating process to withstand elevated temperatures up to 620ºC (1150ºF). The HTC coating can withstand exposure to elevated temperatures without compromising their structural integrity or performance. This is particularly useful in applications where the tweezers may be used in high-temperature environments, such as soldering or other industrial processes. The HTC coating provides enhanced corrosion resistance, protecting the tweezers from damage caused by exposure to heat and potentially corrosive substances. The coating also contributes to the overall durability of the tweezers, ensuring they maintain their functionality and appearance over time, even when subjected to high-temperature conditions. You get increased wear resistance, extending the lifespan of the tweezers and reducing the frequency of replacements. These tweezers are 11 cm ( 4.75”) long.
Catalog No. 50-253-9326