Learn More
HTDialysis LLC HTD Universal Stainless Steel Connecting Rods
Catalog No. NC1696807
Encompass
HTD Universal Stainless Steel Connecting Rods. Replacement part for HTD reusable micro equilibrium dialysis or buffer exchange desalting devices. The HTD devices were developed at a Global Pharmaceutical Research Laboratory with and combine industry-leading accuracy and precision with an automation compatible design for a scalable, cost-efficient, and eco-friendly solutionFor use with:HTD96B – equilibrium dialysis device HTD96C – equilibrium dialysis deviceHTD12A – buffer exchange and desalting device
Catalog No. NC1696807 Supplier HTDialysis LLC Supplier No. 1005
HTD Universal Stainless Steel Connecting Rods. Replacement part for HTD reusable micro equilibrium dialysis or buffer exchange desalting devices. The HTD devices were developed at a Global Pharmaceutical Research Laboratory with and combine industry-leading accuracy and precision with an automation compatible design for a scalable, cost-efficient, and eco-friendly solutionFor use with:HTD96B – equilibrium dialysis device HTD96C – equilibrium dialysis deviceHTD12A – buffer exchange and desalting device