Learn More
HTDialysis LLC HTD Universal Adhesive Sealing Film
Catalog No. NC9107493
Encompass
Adhesive sealing film compatible with all HTD devices. Film is applied to the top surface of the 100% Teflon block during incubation to prevent evaporation and help maintain the proper pH. The sealing film can be removed to access the dialysis samples.
Catalog No. NC9107493 Supplier HTDialysis LLC Supplier No. 1102
Adhesive sealing film compatible with all HTD devices. Film is applied to the top surface of the 100% Teflon block during incubation to prevent evaporation and help maintain the proper pH. The sealing film can be removed to access the dialysis samples.