Learn More
HTDialysis LLC 10kDa MWCO regenerated cellulose dialysis membrane strips. Package contains 8 hydrated individual strips for up to 96 assays. Designed for HTD reusable micro equilibrium dialysis or buffer exchange and desalting devices.
Catalog No. NC2125625
Encompass
Custom cut regenerated cellulose dialysis membrane strips. Designed for HTD reusable micro equilibrium dialysis or desalting devices Features- Range of pore sizes: 1kDa, 3.5kDa, 6-8kDa, 10kDa, 12-14kDa, 25kDa, 50kDa Applications- Serum/plasma protein binding- Microsome protein binding- Relative binding of drug to specific tissues- Receptor binding- T3/T4 free fraction binding (hyperthyroidism) determinations- All desalting and buffer exchange steps of sample processing applications: dialysis of proteins, oligonucleotides, RNA or DNA samples- Removal of sample contaminates - salts, solvents, detergents- Conducting Formation studies - protein-protein, protein-DNA, protein-RNAFor use with:HTD96B – equilibrium dialysis device HTD96C – equilibrium dialysis deviceHTD12A – Buffer exchange and desalting device
This item is not returnable. View return policy
Catalog No. NC2125625 Supplier HTDialysis LLC Supplier No. 1105
Custom cut regenerated cellulose dialysis membrane strips. Designed for HTD reusable micro equilibrium dialysis or desalting devices Features- Range of pore sizes: 1kDa, 3.5kDa, 6-8kDa, 10kDa, 12-14kDa, 25kDa, 50kDa Applications- Serum/plasma protein binding- Microsome protein binding- Relative binding of drug to specific tissues- Receptor binding- T3/T4 free fraction binding (hyperthyroidism) determinations- All desalting and buffer exchange steps of sample processing applications: dialysis of proteins, oligonucleotides, RNA or DNA samples- Removal of sample contaminates - salts, solvents, detergents- Conducting Formation studies - protein-protein, protein-DNA, protein-RNAFor use with:HTD96B – equilibrium dialysis device HTD96C – equilibrium dialysis deviceHTD12A – Buffer exchange and desalting device