Learn More
HTDialysis LLC 1kDa MWCO regenerated cellulose dialysis membrane strips. Package contains 8 hydrated individual strips for up to 96 assays. Designed for HTD reusable micro equilibrium dialysis or buffer exchange and desalting devices.
Supplier:  HTDialysis LLC 1151
Encompass
Custom cut regenerated cellulose dialysis membrane strips. Designed for HTD reusable micro equilibrium dialysis or desalting devices Features- Range of pore sizes: 1kDa, 3.5kDa, 6-8kDa, 10kDa, 12-14kDa, 25kDa, 50kDa Applications- Serum/plasma protein binding- Microsome protein binding- Relative binding of drug to specific tissues- Receptor binding- T3/T4 free fraction binding (hyperthyroidism) determinations- All desalting and buffer exchange steps of sample processing applications: dialysis of proteins, oligonucleotides, RNA or DNA samples- Removal of sample contaminates - salts, solvents, detergents- Conducting Formation studies - protein-protein, protein-DNA, protein-RNAFor use with:HTD96B – equilibrium dialysis device HTD96C – equilibrium dialysis deviceHTD12A – Buffer exchange and desalting device
Catalog No. NC2125623