Learn More
HTDialysis LLC HTD 96B - Reusable 96 Well Equilibrium Dialysis Device - Complete Unit
Supplier:  HTDialysis LLC 1006
Encompass
The HTD96B micro equilibrium device was developed at a Global Pharmaceutical Research laboratory to assess serum protein binding, microsomal protein binding, and tissue specific binding of drug candidates, conserving sample quantities and achieving rapid equilibriumReusable device combines industry-leading accuracy and precision with an automation compatible design for a scalable, cost-efficient, and eco-friendly solutionFeatures• Simultaneously process 1-96 assays• Minimal non-specific binding due to 100% Teflon block construction• Ready in minutes and achieves equilibrium in 4-6 hours on average• Conserves valuable reagents with working volumes of 30-150 ul• Automatable with standard robotic workstations and 96-well pipettors• Ready access to both the sample and dialysate sides of the membrane from the top of the device• Compatible with a wide range of dialysis membrane pore sizes: 1kDa, 3.5kDa, 6-8kDa, 10kDa, 12-14kDa, 25kDa, 50kDa
Catalog No. NC9181256
Add to Cart
Request a Quote